Learn More
Hach Company Free Copper Reagent Powder Pillows, 25 mL, pk/100
Supplier:  Hach Company 2118669
Encompass
For free (un-chelated) copper determination in salt and fresh water by the bicinchoninate method. Sample Size: 25 mL. Pack of 100 Powder Pillows.
Catalog No. 50-199-5443