Learn More
Electron Microscopy Sciences Copper Shot 50 GR
SDP
Supplier:  Electron Microscopy Sciences 2620103
Encompass_Preferred
Used in Canaliculi and Lacunae Stain, Powers, Rasmussen and Clark (1951) for bones and teeth. See Powers, M.M., Rasmusen, G.L., and Clark, G.,Anat. Rec., 111:117- 6, (1951), Clark, G. (ed), Staining Procedures, 3rd edition, William & Wilkins, Baltimore, c 1973, p. 136, Bodian, D., Anat. Rec., 65:89 – 97, 1936, or Bodian, D., Anat. Rec., 69:153 – 62, 1937
Catalog No. 50-317-19