Learn More
Diagnostic Biosystems COPPER STAIN Copper Stain
Supplier:  Diagnostic Biosystems KT033
Encompass_Preferred
The Copper Stain Kit (For Microwave) is intended for the demonstration of copper deposits in tissue sections.
Catalog No. 50-240-7507