Learn More
Medchemexpress LLC Copper sulfate pentahydrate | 7758-99-8 | MFCD00149681 | 99.9% | 500mg
SDP
Supplier:  Medchemexpress LLC HYY1881A500MG
Encompass_Preferred
Copper sulfate pentahydrate 99% is a biochemical reagent Copper sulfate pentahydrate 99% reduces the production of ROS and the expression levels of MyD88 as well as c-Rel genes Copper sulfate pentahydrate 99% decreases the activities of T-SOD CAT and GSH increases the activities of caspase-3 caspase-8 and caspase-9 Copper sulfate pentahydrate 99% is cytotoxic to various cells Copper sulfate pentahydrate 99% has antioxidant activity Copper sulfate pentahydrate 99% can be used in the research of diabetes Parkinson s disease and DMBA (HY-W011845)-induced tumors[1][2][3][4][5][6][7]
Catalog No. 50-002-65954