Learn More
Medchemexpress LLC Copper tripeptide-3 | 767286-83-9 | 99.09% | 415.93 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYP5243
Encompass_Preferred
Copper tripeptide-3 (AHK-Cu) is a bioactive peptide known for its hair growth promotion effects. It stimulates dermal cell proliferation and viability, increasing collagen deposition to renew the extracellular matrix. This peptide also encourages the elongation of human hair follicles and the proliferation of dermal papilla cells.
  • Increases growth and viability of dermal fibroblasts
  • Stimulates collagen production
  • Promotes proliferation of dermal papilla cells
  • Reduces apoptotic dermal papilla cells
  • Increases Bcl-2 expression and decreases Bax expression
Catalog No. 50-003-98791