Learn More
Medchemexpress LLC Copper tripeptide-3 (AHK-Cu) | 767286-83-9 | 99.1% | 415.93 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYP5243100MG
Encompass_Preferred
Copper tripeptide-3 (AHK-Cu) is a bioactive peptide that promotes hair growth. It enhances dermal cell proliferation and viability, and increases collagen deposition to regenerate the extracellular matrix. It also stimulates the elongation of human hair follicles and the proliferation of dermal papilla cells.
  • Increases the growth and viability of dermal fibroblasts and stimulates collagen production in normal human dermal fibroblasts.
  • Significantly stimulates the proliferation of cultured dermal papilla cells.
  • Reduces the number of apoptotic dermal papilla cells.
  • Increases Bcl-2 expression and decreases Bax expression in dermal papilla cells.
Catalog No. 50-003-96393