Learn More
Sigma Aldrich Fine Chemicals Biosciences CopperII sulfate puriss. p500G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 61230500GF
Encompass
Copper(II) sulfate (CuSO4) is an anhydrous copper(II) salt. It can be synthesized from CuSO4.5H2O via dehydration. Copper(II) sulfate adsorbed on silica gel participates in the conversion of secondary and tertiary alcohols to olefins. Copper(II) sulfate-alumina has been employed as a solid-reagent system during the synthesis of benzils.
Catalog No. 50-289-9711