Learn More
Medchemexpress LLC Coproporphyrin III | 14643-66-4 | MFCD06804602 | 99.6% | 654.71 g/mol | C36H38N4O8 | 1 ML

Supplier: Medchemexpress LLC HY10139810MM 1ML
Coproporphyrin III (Zincphyrin) is an endogenous porphyrin metabolite provided as a 10 mM solution in DMSO for research use. It is suitable as a biochemical reagent or reference standard in metabolic and porphyrin-related studies.
- Supplied as a 10 mM solution in DMSO (1 mL).
- High purity (>99.5%).
- Molecular formula C36H38N4O8; molecular weight 654.71 g/mol.
- Soluble in DMSO; in vitro solubility reported at 125 mg/mL (ultrasonic).
- In vivo formulation protocols and solubility guidance available.
- Datasheet and certificate of analysis available to support use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.