Learn More
Selleck Chemical LLC Coptisine chloride S5249-25mg
SDP
Supplier:  Selleck Chemical LLC S524925MG
Encompass_Preferred
Coptisine (Q-100696 NSC-119754) is an isoquinoline alkaloid isolated from Coptidis Rhizoma with anti-diabetic antimicrobial antiviral anti-hepatoma and anti-leukemia effects Coptisine chloride is an efficient uncompetitive IDO inhibitor with IC50 value of 6 3 M and Ki value of 5 8 M
Catalog No. 50-311-1921