Learn More
LGC GENOMICS LLC (Lucigen) CopyCutter Induction Solution, 25 mLInduce plasmid copy number in CopyCutter EPI400 strain, for high plasmid yields.
Supplier:  LGC GENOMICS LLC CIS40025
Encompass
Prepare unbiased fosmid libraries of approx. 40 kb that can be maintained at single-copy number and induced to high-copy number as needed when grown in the TransforMax EC300 E. coli.
Catalog No. NC1419826