Learn More
Grainger Cor Rad End Mill 1/2 Carb 0.0200 rad
Supplier:  Grainger 14N082
Encompass_Preferred
The Corner Radius End Mill from Grainger is a 4-flute carbide end mill designed for corner radiusing and contouring in a variety of materials. It features a 30 degree helix angle and right-hand cut. The end mill has a neck relief to reduce chatter and improve finish. It is available in multiple corner radius sizes from 1/64" to 1/4", and shank diameters of 1/8", 3/16", 1/4", 5/16", 3/8", and 1/2". The end mill is made in the USA out of micrograin carbide for wear resistance and heat tolerance. Its applications include die/mold work, profiling, slotting, and contouring in materials like tool steels, hardened steels, titanium alloys, nickel alloys, and stainless.
Catalog No. 50-275-2082