Learn More
Grainger Cor Rad End Mill 1.50mm Carb 0.15mm rad
Supplier:  Grainger 14K431
Encompass_Preferred
The Micro 100 Ball End Mill is a 4 flute carbide end mill with a ball end. It has a 1/4" shank diameter and is available in multiple diameters from 1/8" to 1/2". The ball end allows milling of 3D contoured surfaces. Carbide provides wear resistance and tool life. The 4 flute design provides better chip evacuation than a 2 flute. This end mill is well suited for machining complex 3D contours in materials like steel, aluminum, and plastics. It can be run at speeds up to 25,000 RPM. Overall, this Micro 100 ball end mill is a versatile tool for 3D contouring applications.
Catalog No. 50-275-3369