Learn More
Grainger Cor Rad End Mill 3.00mm Carb 0.50mm rad
Supplier:  Grainger 14T391
Encompass_Preferred
The Micro 100 Corner Radius End Mill is a 4-flute carbide end mill with a corner radius of 0.031 inches. It has a 1/4 inch shank diameter and is available in multiple overall lengths from 1/2 inch to 2 inches. The end mill has a TiAIN coating and can machine a variety of materials including steel, stainless steel, cast iron, aluminum, copper, brass, bronze, plastic and graphite. It is designed for high feed milling applications. The end mill provides accurate corner radii and can mill small pockets, slots and contours.
Catalog No. 50-275-1580