Learn More
Grainger Cor Rad End Mill 3/16 Carb 0.0200 rad
Supplier:  Grainger 14T007
Encompass_Preferred
The Micro 100 Corner Radius End Mill with AlTiN coating is a high performance end mill designed for machining aluminum, titanium, and other non-ferrous metals. It features a 0.125" corner radius and 0.25" shank diameter, with 2 flutes and a 14 degree helix angle. The AlTiN coating provides heat and wear resistance. Overall, this is an end mill optimized for efficient machining and extended tool life in non-ferrous metals.
Catalog No. 50-275-0208