Learn More
Grainger Cor Rad End Mill 3/8 Carb 0.0100 rad
Supplier:  Grainger 14T069
Encompass_Preferred
The Micro 100 Corner Radius End Mill is a bright finish, carbide end mill with a corner radius of 0.031 inches. It has a 1/4 inch shank diameter and is available in a 2 flute design. The overall length is 2 inches and it can be run at speeds up to 25,000 RPM. This end mill is made of solid carbide and can machine a variety of materials including steel, stainless steel, cast iron, aluminum, copper, brass, bronze, plastic and graphite. It features a TiAlN coating for increased tool life and lubricity. The corner radius design allows for strong cutting edges and smooth wall blends. This is an ideal tool for slotting, side milling and profiling operations.
Catalog No. 50-275-1682