Learn More
Grainger Cor Rad End Mill 3/8 Carb 0.0200 rad
Supplier:  Grainger 14N078
Encompass_Preferred
The WIDIA HANITA 785WF8 is a corner radius end mill made of micrograin carbide with an AlTiN coating. It has a 0.5" cutter diameter, 0.125" corner radius, 2 flute design, and 0.63" overall length. This end mill is designed for high feed milling of steel, stainless steel, hardened steel, titanium, nickel based alloys, and other materials. It provides good wear resistance and heat resistance. The AlTiN coating improves tool life in high heat applications. This is a high performance end mill well-suited for difficult to machine materials.
Catalog No. 50-275-2086