Learn More
Grainger Cor Rad End Mill 5/16 Carb 0.0100 rad
Supplier:  Grainger 14T048
Encompass_Preferred
The Micro 100 Corner Radius End Mill is a bright finish end mill made of solid carbide with a corner radius of 0.031 inches. It has 2 flutes and a right hand spiral. The overall length is 2 inches and shank diameter is 0.25 inches. It is designed for general purpose milling in steel, stainless steel, cast iron, aluminum, and other non-ferrous materials. The end mill features a TiAlN coating for increased tool life and heat resistance. It provides good chip evacuation while milling and can be run at speeds up to 21,000 RPM. The corner radius profile allows milling fillets and contours.
Catalog No. 50-275-1791