Learn More
Grainger Cor Rad End Mill 5/16 Carb 0.0100 rad
Supplier:  Grainger 14T035
Encompass_Preferred
The Micro 100 Corner Radius End Mill from Grainger is a bright finish, carbide end mill with a corner radius of 0.031 inches. It has a 1/4 inch shank diameter and a 1/2 inch overall length. The end mill has 2 flutes and can be run at speeds up to 25,000 RPM with feed rates up to 80 IPM. It is made in the USA and well-suited for machining aluminum, brass, bronze, copper, plastics, and other non-ferrous materials. This versatile end mill can be used for slotting, side milling, and contouring operations.
Catalog No. 50-275-0510