Learn More
Grainger Cor Rad End Mill 8.00mm Carb 0.50mm rad
Supplier:  Grainger 14M985
Encompass_Preferred
The Micro 100 Corner Radius End Mill is a carbide end mill with a corner radius of 0.031 inches. It has a 1/4 inch shank diameter and is available in multiple lengths from 1/2 inch to 2 inches. The end mill has 4 flutes and can be run at speeds up to 12,000 RPM. It is designed for general purpose milling in steels, stainless steels, cast iron, aluminum and other non-ferrous materials. The end mill features TiAlN coating for increased tool life and performance. It produces a smooth corner radius on the workpiece.
Catalog No. 50-274-9695