Learn More
Medchemexpress LLC HY-137204 5mg Medchemexpress, COR659 CAS:544450-68-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1372045MG
Encompass_Preferred
Medchemexpress, HY-137204 5mg COR659 CAS:544450-68-2 COR659 is a potent and effective GABAB positive allosteric modulator (PAM). COR659 suppresses alcohol and chocolate self-administration in rats. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1475