Learn More
Proteintech Group Inc CoraLite488 TUNEL Assay Apoptosis Detection Kit(green)
SDP
Supplier:  Proteintech Group Inc PF0000620T
Encompass_Preferred
A distinctive feature of apoptosis is degradation of DNA. The DNA fragments tend to be vary in length in integer multiples of 180 bp-200 bp, which is shown as a ladder-like pattern in agarose gel electrophoresis. This kit uses the TUNEL method, using Terminal Deoxynucleotidyl Transferase (TdT) to catalyze the incorporation of CL488-dUTP at the 3'-OH end of the broken DNA of apoptotic cells. CL488-dUTP-labeled DNA can be directly observed with a fluorescence microscope or quantified with a flow cytometer. The TUNEL method can selectively detect apoptotic cells, but not necrotic cells or cells with DNA strand breaks caused by irradiation and drug treatment.
Catalog No. 50-225-8712