Learn More
Medchemexpress LLC Coralyne chloride | 38989-38-7 | 98.3% | 399.87 g/mol | C22H22ClNO4 | 100 MG

Supplier: Medchemexpress LLC HY118581100MG
Coralyne chloride is the chloride salt of the protoberberine alkaloid coralyne, provided as a solid research reagent with reported topoisomerase I-poison activity and anti-cancer properties.
- Supplied as a 100 mg solid pack for research use.
- High reported purity suitable for biochemical assays.
- Soluble in water and DMSO with heating and sonication.
- Stable when stored sealed at 4°C; solution storage guidelines provided.
- Molecular weight and formula useful for formulation and dosing calculations.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.