Learn More
Medchemexpress LLC Coralyne chloride | 38989-38-7 | MFCD00011938 | 98.3% | 399.87 g/mol | C22H22ClNO4 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY11858125MG
Encompass_Preferred
Coralyne chloride is a protoberberine alkaloid supplied as the chloride salt for laboratory research. It binds nucleic acids, acts as a topoisomerase I poison, and is used in biochemical and anticancer studies; supplied as a solid at high purity for in vitro applications.
  • Protoberberine alkaloid with DNA-binding properties.
  • Topoisomerase I poison used in anticancer and biochemical studies.
  • Available as a chloride salt solid suitable for in vitro research.
  • High purity (98.3%) for analytical and biological assays.
  • Multiple pack sizes, including 25 MG for small-scale experiments.
  • Useful as a fluorescent probe in DNA interaction studies.
Catalog No. 50-002-26444