Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Coralyne chloride hydrate | 38989-38-7 | MFCD00011938 | 98.3% | 399.87 g/mol | C22H22ClNO4 | 50 MG
SDP

Supplier:  Medchemexpress LLC HY11858150MG

Encompass_Preferred

Coralyne chloride is a protoberberine alkaloid research reagent that acts as a topoisomerase I poison and induces Top I-mediated DNA cleavage. It is used in anticancer research and as a precursor for DNA-binding fluorescent probes. The compound is supplied at high purity and characterized for solubility and storage conditions.

  • Purity: 98.3%.
  • Molecular weight: 399.87 g/mol.
  • Chemical formula: C22H22ClNO4.
  • Solubility: H2O 4 mg/mL; DMSO 1.82 mg/mL (heat and ultrasonic recommended).
  • Storage: store sealed at 4°C; stock solutions -80°C (6 months) or -20°C (1 month).
  • Applications: topoisomerase I studies and fluorescent DNA probe synthesis.

Catalog No. 50-002-27326


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.