Learn More
Kipper Tool Company CORDLESS ELEC RING CUTTER
SDP
Supplier:  Kipper Tool Company GRO 48.0610
Encompass_Preferred
Cordless battery powered ring cutter zips through most finger rings in hospital emergency rooms in less than 10 seconds. Finger guard stays between the cutting blade and the patient's finger at all times. This protects the patient and makes it easy and safe for hospital staff to remove the ring. Patented design conducts heat away from the ring and the finger. No need for external irrigation or frequent stops. The patient is comfortable. Ring is removed. Staff is free to handle the next emergency. Will cut platinum and titanium rings, depending upon grade, in a longer timeframe, with stops. Tungsten rings will damage this ring cutter. Chrome blade assembly can be sterilized
Catalog No. NC9020117