Learn More
CORDX LABS INC CorDx Tyfast Flu A/B & COVID-19 Multiplex Rapid Test (10PK/CS)
Supplier:  CORDX LABS INC ACT2100110
Encompass_Preferred
"The CorDx Tyfast Flu A/B & COVID-19 Multiplex Rapid Test is a lateral flow immunoassay intended for the qualitative and differential detection of SARS-CoV-2 and Influenza A and B antigen in an anterior nares sample (from both nostrils). The test provides an efficient and accurate diagnostic solution, giving communities and families fast way to prioritize their health.
CPT code: COVID-19: 87811QW   
Flu A: 87804QW     
Flu B: 87804QW-59
Features and Benefits 
The First 10-minute test in the US for Flu A/B & COVID-19. 
No instrument needed.
High sensitivity, specificity and accuracy.
Only need 3 drops of solution.
Evaluated against XBB variants
Storage: 36-86°F/2~30°C. 
Shelf Life: 15 months
CorDx Tyfast Emergency Use Authorization Number(EUA): EUA230055
This product has not been FDA cleared or approved but has been authorized by FDA under an EUA for us by authorized laboratories.
Catalog No. NC3363552