Learn More
MALIX LLC CorDx COVID-19 Ag Test, 2 Pack
Supplier:  MALIX LLC CORDX_OTC2PK
Encompass_Preferred
Minimum Order Quantity: Must Order by Case quantity (150 Kits)
The CorDx COVID-19 Ag Test is a lateral flow immunoassay device intended for the qualitativedetection of nucleocapsid protein antigen from the SARS-CoV-2 virus.
Catalog No. NC2480792