Learn More
Cayman Chemical Cordyceps mIlItarIs 10mg
SDP
Supplier:  Cayman Chemical 1442610 MG
Encompass_Preferred
A nucleoside analog with diverse biological activities; is converted to cordycepin triphosphate, which inhibits ATP-dependent DNA and RNA synthesis; incorporates into DNA and RNA in SK-HEP-1 cells; is active against B. subtilis at 0.1-1 mg/ml; increases IL-10 secretion and mRNA expression in isolated human PBMCs at 24 UG/ml; increases survival of mice infected with C. albicans or C. krusei at 1 mg/kg per day
Catalog No. 50-297-0425