Learn More
Medchemexpress LLC (-)-Corey lactone diol | 32233-40-2 | 172.18 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW008393
Encompass_Preferred
(-)-Corey lactone diol is the reduced form of Corey aldehyde. It can be used as a drug intermediate for the synthesis of Beraprost.
  • Reduced form of Corey aldehyde
  • Can be used as a drug intermediate
  • Building block in chemical synthesis
Catalog No. 50-004-49445