Learn More
Medchemexpress LLC (-)-Corey lactone diol | 32233-40-2 | 172.18 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0083935G
Encompass_Preferred
(-)-Corey lactone diol is the reduced form of Corey aldehyde. It is used as a drug intermediate for the synthesis of Beraprost.
  • Drug intermediate
  • Chiral building block
  • Used in the synthesis of prostaglandin analogue Beraprost
  • Relevant for inflammation or immune system disease research
  • Relevant for blood or cardio-cerebrovascular disease research
Catalog No. 50-004-45577