Learn More
Medchemexpress LLC HY-N0462 5mg Medchemexpress, Corilagin CAS:23094-69-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN04625MG
Encompass_Preferred
Medchemexpress, HY-N0462 5mg Corilagin CAS:23094-69-1 Corilagin, a gallotannin, is isolated from Caesalpinia coriaria (Jacq.) Willd. Corilagin inhibits activity of reverse transcriptase of RNA tumor viruses. Corilagin inhibits the growth of Staphylococcus aureus with a MIC of 25 μg/mL. Corilagin shows good anti-tumor activity on hepatocellular carcinoma and ovarian cancer. Corilagin shows a low level of toxicity toward normal cells and tissues. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0275