Learn More
Selleck Chemical LLC Corilagin S9090-1MG
SDP
Supplier:  Selleck Chemical LLC S90901MG
Encompass_Preferred
Corilagin discovered in many medicinal plants such as Phyllanthus speices etc has multiple activities including anti-oxidative anti-inflammatory anti-apoptotic hepatoprotective and others It is an inhibitor of TNF-
Catalog No. 50-000-25045