Learn More
Medchemexpress LLC Corin | 1808113-09-8 | 98.2% | 428.53 | 100 MG

Supplier: Medchemexpress LLC HY11104810MM/1ML
Corin is a dual inhibitor targeting histone lysine specific demethylase (LSD1) and histone deacetylase (HDAC), with a Ki(inact) of 110 nM for LSD1 and an IC50 of 147 nM for HDAC1. It is a chemical reagent intended for research use only and not for human use.
- Inhibits the deacetylation of semisynthetic, reconstituted nucleosomes by the CoREST ternary complex.
- Demonstrates irreversible inhibition of HDAC1 activity.
- More potent and efficacious in inducing cellular H3K9 acetylation compared to MS-275.
- Non-toxic to primary human melanocytes at 1 μM concentration.
- Available in various quantities and forms including solid or 10 mM solution.
- Shipping at room temperature.
- Storage recommendations include -20°C under nitrogen for solid form and -80°C or -20°C in solvent.
- Solubility in DMSO is 100 mg/mL.
- Protocols are provided for preparing in vivo working solutions.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.