Learn More
Julabo USA Inc CD-B13 heating bath circulator with stainless steel bath tank, 115V/560Hz, 20…150C range, 1kW heater, 13L SS tank, Pump: 6 L/min, 1.5 PSI. CSA/UL marked.
Supplier: Julabo USA Inc 9012413.02
Heating bath circulator with stainless steel bath tank, CD-B13, 115V/60Hz, 20…150C. 1kW heater. Pressure centrifugal pump 15LPM, 1.5 PSI. 17...27L bath tank. Bath tank opening: 11.8x7.1/8.9 in (WxL/D); Dimensions: 15 x 15.7x 16.5 in. 20.1lbs. M16x1 threaded pump connections with 8mm and 12mm hose barbs. USB-A data-recording; USB-B external control. CSA/UL Marked.