Learn More
Julabo USA Inc CD-BC12 heating circulator, 115V/60Hz, 20…150C. 1kW heater. 12L tank. CSA/UL
Supplier: Julabo USA Inc 9012512.02
Heating circulator, CD-BC12, 115V/60Hz, 20…150C. 1kW heater. Pump to 17LPM to 6.2 PSI. USB-A data-logging; USB-B external communication. M16x1 pump fittings with 8 and 12mm barbs. 8.5…12L bath tank. Bath opening; 8.7x5.9/7.9 in (WxL/D), Dimensions: 13 x 19.3 x 18.5 in. 27lbs. CSA/UL marked