Learn More
Selleck Chemical LLC CORM-3
SDP
Supplier:  Selleck Chemical LLC S74481G
Encompass_Preferred
CORM-3 is a carbon monoxide-releasing molecule with anti-inflammatory and cardioprotective activity
Catalog No. 50-000-01366