Learn More
Sigma Aldrich Fine Chemicals Biosciences Corn oil, 8001-30-7, MFCD00130865, 500 mL
Supplier:  Sigma Aldrich Fine Chemicals Biosciences C8267500ML
Encompass
Delivery vehicle for fat-soluble compounds, Synonym: Maize oil. Corn oil has been used in research applications as delivery vehicle for fat-soluble compounds including toxins. It is also used in the delivery of tamoxifen (TAM) in mice, curcumin in rats and [3H]retinol in mice. Corn oil has been used in research applications as delivery vehicle for fat-soluble compounds including toxins.
Catalog No. 50-178-0915