Learn More
ATCC Corneal Epithelial Cell Growth Kit
Supplier:  ATCC PCS700040
Encompass
Corneal Epithelial Cell Growth Kit (ATCC PCS-700-040) contains components that when added to Corneal Epithelial Cell Basal Medium (ATCC PCS-700-030) create a complete ATCC Primary Cell Solutions culture environment for corneal epithelial cells derived from a normal human cornea (e.g., Primary Corneal Epithelial Cells, Normal, Human, ATCC PCS-700-010). The serum-free medium formulation is designed to support normal corneal cell morphology, and promote rapid growth, and proliferation. No feeder layers, extracellular matrix proteins, or other substrates are required.
Catalog No. 50-238-1701