Learn More
Visikol Inc 2 mm Coronal Mouse Brain Slicer
Supplier:  Visikol Inc BSLC1
Encompass_Preferred
Histological Accessory, Tissue Preperation Accessory, 2 mm Coronal Mouse Brain Slicer, 2mm. The 2 mm Coronal Mouse Brain Slicer allows you to easily cut a brain into smaller pieces for easier imaging with objectives that have a limited working distance. This mouse brain slicer is 3D printed out of PLA and can be easily washed and cleaned between use. All you need is a standard razor blade to start slicing your tissues.
Catalog No. 50-161-6426