Learn More
Medchemexpress LLC Coronastat | 2922281-15-8 | MFCD32757276 | 98.0% | 575.53 g/mol | C22H29F3N3NaO8S | 5 MG

Supplier: Medchemexpress LLC HY1470205MG
Coronastat is a research-grade small-molecule inhibitor of the SARS-CoV-2 3CL protease intended for in vitro and in vivo pharmacology studies. The compound has molecular weight 575.53 g/mol and CAS 2922281-15-8; manufacturer datasheet provides solubility and storage recommendations for experimental use.
- Potent inhibitor of the SARS-CoV-2 3CL protease.
- High reported purity of 98.0%.
- Soluble in DMSO and water at high in vitro concentrations.
- In vivo formulations achieve ≥ 2.08 mg/mL solubility in recommended vehicles.
- Storage: 4°C sealed; in solvent -80°C (6 months) or -20°C (1 month).
- Supplied in small quantities suitable for assay development and dosing studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.