Learn More
Selleck Chemical LLC Coronastat-E1174-5MG
SDP
Supplier:  Selleck Chemical LLC E11745MG
Encompass_Preferred
Coronastat is a potent inhibitor of the SARS-CoV-2 3CL protease which is a critical drug target for small molecule COVID-19 given its likely druggability and essentiality in the viral maturation and replication cycle
Catalog No. 50-313-5703