Learn More
Quanterix Inc CORPLEX HUMAN CYTOKINE PANEL 1
SDP
Supplier:  Quanterix Inc 850329
Encompass
(ifn-g,il-1b,il-4,il-5,il-6,il-8,il-10,il-12p70,il-22,tnfa)includes planar array plate, calibrators, diluents, biotinylated detector antibody mix, wash buffer, streptavidin-hrp, and chemiluminescent substrate
Catalog No. NC1828119