Learn More
Thermo Scientific CORROSION RESISTANT VACUUM PU
Supplier:  Thermo Scientific VLP80115
Encompass_Preferred
Pump; Oil Vacuum; Thermo Scientific; VLP120 series; Displacement: 76L/min (at 60Hz), 62L/min (at 50Hz); 6 x 17 x 9 in. (160 x 420 x 230mm); Ultimate Vacuum Level 1.5 mtorr/1.95 x10minus3 mbar; 115V 60Hz; 0.7L
Catalog No. 50-870-639