Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Value Collection Corrugated Sheet, 28 x 22 in, 50/Pack

Supplier:  MSC 39592696

Encompass

This corrugated sheet provides robust protection for various packaging, shipping, and layering applications. Its durable corrugated construction offers cushioning and rigidity for items requiring safeguarding during transit or storage.

  • Long: 28 in
  • Wide: 22 in
  • Origin: Made in USA
  • Available in multiples of 50

Catalog No. 50-191-9653


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.