Learn More
MSC MSC Corrugated Sheet, Kraft, 24 x 36 in, 5 sheets
Supplier:  MSC 39592738
Encompass
Corrugated sheets are designed for use as layering pads on pallets, helping to disperse weight and prevent damage during transport and storage. They also serve to keep products dust-free.
  • Disperses weight and prevents strapping and forklift damage
  • Keeps products dust-free during storage
  • Made in USA
  • Sold in bundle quantities
Catalog No. 50-191-9657