Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Generic Corrugated Sheet, 36 x 24 in, White, 5 sheets

Supplier:  MSC 39575899

Encompass

This corrugated sheet is designed for various industrial applications, including layering on pallets to disperse weight and protect goods from dust and damage during storage and transit. It is constructed from durable 200#/ECT-32 white corrugated material.

  • Made in USA
  • Ideal for layering pads on pallets
  • Helps keep product tops dust-free
  • Prevents strapping and forklift damage
  • White on one side
  • Dimensions: 36 in long x 24 in wide
  • Box quantity: 5 sheets

Catalog No. 50-191-8004


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.