Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Partners Brand Corrugated Sheets, 36 x 96 in., Kraft, 5/Bundle

Supplier:  MSC 39592803

Encompass

These corrugated sheets provide versatile packaging and protection. Made from durable single-wall corrugated material, they are suitable for various industrial and commercial applications.

  • Dimensions: 36 in. width x 96 in. length
  • Material: 200#/ECT-32 single wall corrugated
  • Color: Kraft
  • Package quantity: 5 sheets per bundle

Catalog No. 50-191-9664


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.