Learn More
TARGETMOL CHEMICALS INC CORT108297 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T150005MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Cort108297 is a selective glucocorticoid receptor modulator and a selective GR antagonist with no affinity for other steroid receptors. Cort108297 showed a high affinity for GRs (Ki 0.45nM). purity: 98%
Catalog No. 50-258-1262