Learn More
Waters Corp CORTECS UPLC C18 Column, 90Å, 1.6 µm, 2.1 mm X 100 mm, 3/pk
Supplier:  Waters Corp 176003149
Encompass_Preferred
CORTECS C18 Columns are general purpose, high-efficiency columns based on a solid-core particle that offer balanced retention of acids, bases and neutrals at low and mid-range pH.
Catalog No. 50-597-035