Learn More
Waters Corp Waters Chromatography Column, C18+, 2.7 μm, 2.1 mm x 50 mm
Supplier:  Waters Corp 186007395
Encompass_Preferred
CORTECS C18+ columns are general purpose, charged surface, high efficiency C18 columns based on a solid-core particle designed to deliver excellent peak shape for basic analytes at low pH. The C18 ligand is chosen for its enhanced data retention and stability capabilities. It allows for resolving complicated mixes or maintaining resolution while improving throughput.
  • General purpose, charged surface, high efficiency C18 columns based on a solid-core particle.
  • Designed to deliver excellent peak shape for basic analytes at low pH.
  • Enables resolution of complicated mixes or maintains resolution while improving throughput.
  • Solid-core particles enhance performance by reducing terms in the van Deemter equation.
  • Withstands temperatures up to 45°C and functions between 2 and 8 pH.
Catalog No. 50-592-628